missing translation for 'onlineSavingsMsg'
Learn More

(2-Iodoethyl)benzene, 97%, stabilized with copper, Thermo Scientific Chemicals

Catalog Number p-148982
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
17376-04-4
C8H9I
232.07
MFCD00039408
KVTHPKXDLVYNCH-UHFFFAOYSA-N
2-iodoethyl benzene, benzene, 2-iodoethyl, 2-phenylethyl iodide, phenethyl iodide, iodoethyl benzene, .beta.-phenethyl iodide, phenethyliodide, p-iodoethylbenzene, phenylethyl iodide, 4-iodoethylbenzene
28503
2-iodoethylbenzene
C1=CC=C(C=C1)CCI
This item is not returnable. View return policy
17376-04-4
C8H9I
232.07
MFCD00039408
KVTHPKXDLVYNCH-UHFFFAOYSA-N
2-iodoethyl benzene, benzene, 2-iodoethyl, 2-phenylethyl iodide, phenethyl iodide, iodoethyl benzene, .beta.-phenethyl iodide, phenethyliodide, p-iodoethylbenzene, phenylethyl iodide, 4-iodoethylbenzene
28503
2-iodoethylbenzene
C1=CC=C(C=C1)CCI

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Brown-Red
Density 1.6000g/mL
Boiling Point 122.0°C to 123.0°C (13.0 mmHg)
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2CH2I
Refractive Index 1.6000 to 1.6020
Quantity 5 mL
Specific Gravity 1.6
Solubility Information Solubility in water: insoluble in water
Formula Weight 232.07
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material (2-Iodoethyl)benzene, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'