missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Iodobenzyl alcohol, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
2-Iodobenzyl alcohol was used in the synthesis of substituted seven-membered lactones, 2-[(E)-(1?-iodo-2?-propenyl)]benzyl alcohol2 and 2,3-diphenyl-1-indenone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Iodobenzyl alcohol was used in the synthesis of substituted seven-membered lactones, 2-[(E)-(1′-iodo-2′-propenyl)]benzyl alcohol2 and 2,3-diphenyl-1-indenone.
Solubility
Insoluble in water.
Notes
Light Sensitive, store in dark. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
2-Iodobenzyl alcohol was used in the synthesis of substituted seven-membered lactones, 2-[(E)-(1′-iodo-2′-propenyl)]benzyl alcohol2 and 2,3-diphenyl-1-indenone.
Solubility
Insoluble in water.
Notes
Light Sensitive, store in dark. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Melting Point | 89°C to 92°C |
Quantity | 5 g |
Beilstein | 2079487 |
Sensitivity | Light sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 234.04 |
Percent Purity | 99% |
Chemical Name or Material | 2-Iodobenzyl alcohol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'