missing translation for 'onlineSavingsMsg'
Learn More

2-Iodobenzoic acid, 98+%, Thermo Scientific Chemicals

Catalog Number p-7080843
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
88-67-5
C7H5IO2
248.019
MFCD00002419
CJNZAXGUTKBIHP-UHFFFAOYSA-N
o-iodobenzoic acid, benzoic acid, 2-iodo, iodobenzoic acid, benzoic acid, o-iodo, usaf ek-572, 2-iodo benzoic acid, 2-iodo-benzoic acid, benzoic acid, iodo, kyselina o-jodbenzoova, o-jod-benzoesaeure
6941
CHEBI:287979
2-iodobenzoic acid
C1=CC=C(C(=C1)C(=O)O)I
This item is not returnable. View return policy
88-67-5
C7H5IO2
248.019
MFCD00002419
CJNZAXGUTKBIHP-UHFFFAOYSA-N
o-iodobenzoic acid, benzoic acid, 2-iodo, iodobenzoic acid, benzoic acid, o-iodo, usaf ek-572, 2-iodo benzoic acid, 2-iodo-benzoic acid, benzoic acid, iodo, kyselina o-jodbenzoova, o-jod-benzoesaeure
6941
CHEBI:287979
2-iodobenzoic acid
C1=CC=C(C(=C1)C(=O)O)I

Reagent for detection of sulfhydryl groups in proteins2-Iodobenzoic acid acts as a precursor in the preparation of 2-Iodoxybenzoic acid (IBX) and Dess-Martin periodinane, which finds application as an oxidizing agent in synthetic chemistry. It is also used in Suzuki reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent for detection of sulfhydryl groups in proteins2-Iodobenzoic acid acts as a precursor in the preparation of 2-Iodoxybenzoic acid (IBX) and Dess-Martin periodinane, which finds application as an oxidizing agent in synthetic chemistry. It is also used in Suzuki reaction.

Solubility
Soluble in dimethyl sulfoxide and methanol

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 160°C to 164°C
Density 2.25
Quantity 25 g
Beilstein 1861406
Sensitivity Light sensitive
Merck Index 14,5030
Solubility Information Soluble in dimethyl sulfoxide and methanol
Formula Weight 248.02
Percent Purity ≥98%
Chemical Name or Material 2-Iodobenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'