missing translation for 'onlineSavingsMsg'
Learn More

2-Iodoanisole, 97%, ACROS Organics™

Brand:  Acros Organics 249680500

Code : 29

Additional Details : CAS Number : 529-28-2 Weight : 0.10000kg


Packaging Glass bottle
Quantity 50g

 View more versions of this product

Product Code. 10637592

  • £69.80 / 50g


Estimated Shipment
19-07-2018
Add to basket


Specifications

Chemical Identifiers

2-Iodoanisole
529-28-2
100.0
97%
IC6H4OCH3
MFCD00001039
97%
96.0
≥96.0%
C7H7IO
234.04
1-Iodo-2-methoxybenzene

Specifications

2-Iodoanisole
529-28-2
100.0
Glass bottle
≥96.0%
1.6220 to 1.6240 (20°C, 589nm)
IC6H4OCH3
MFCD00001039
15, 5071
1-Iodo-2-methoxybenzene
50g
97%
96.0
Authentic
Liquid
97%
C7H7IO
234.04
Product may darken on storage
1.79g/mL
Clear yellow
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Iodoanisole

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS