missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-5-methylthiophene, 97%, Thermo Scientific Chemicals

Catalog Number 11956841
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
16494-36-3
C5H5IS
224.06
MFCD00022492
NAZNQEXKAPLVKC-UHFFFAOYSA-N
thiophene, 2-iodo-5-methyl, pubchem7417, acmc-1bs6v, 2-iodo-5-methyl-thiophene, 2-iodo-5-methyl thiophene, bidd:gt0402, 2-iodo-5-methylthiophene
140096
2-iodo-5-methylthiophene
CC1=CC=C(I)S1
This item is not returnable. View return policy
16494-36-3
C5H5IS
224.06
MFCD00022492
NAZNQEXKAPLVKC-UHFFFAOYSA-N
thiophene, 2-iodo-5-methyl, pubchem7417, acmc-1bs6v, 2-iodo-5-methyl-thiophene, 2-iodo-5-methyl thiophene, bidd:gt0402, 2-iodo-5-methylthiophene
140096
2-iodo-5-methylthiophene
CC1=CC=C(I)S1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point -25°C to 28°C
Color Orange-Red to Yellow
Density 1.8520g/mL
Boiling Point 81°C to 83°C (10.0 mmHg)
Flash Point 89°C
Infrared Spectrum Authentic
Assay Percent Range 96.5% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Specific Gravity 1.852
Formula Weight 224.06
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Iodo-5-methylthiophene
Show More Show Less
compliance-icons
Product Identifier
  • 2-Iodo-5-methylthiophene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'