missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-4-nitrotoluene, 97%, Thermo Scientific Chemicals

Catalog Number p-7025809
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
7745-92-8
C7H6INO2
263.03
MFCD00024329
BUQSRXQJUZTIEW-UHFFFAOYSA-N
2-iodo-4-nitrotoluene, benzene, 2-iodo-1-methyl-4-nitro, 2-iodo-1-methyl-4-nitro-benzene, pubchem3957, acmc-209pa2, toluene, 2-iodo-4-nitro, timtec-bb sbb005821
82188
2-iodo-1-methyl-4-nitrobenzene
CC1=CC=C(C=C1I)[N+]([O-])=O
This item is not returnable. View return policy
7745-92-8
C7H6INO2
263.03
MFCD00024329
BUQSRXQJUZTIEW-UHFFFAOYSA-N
2-iodo-4-nitrotoluene, benzene, 2-iodo-1-methyl-4-nitro, 2-iodo-1-methyl-4-nitro-benzene, pubchem3957, acmc-209pa2, toluene, 2-iodo-4-nitro, timtec-bb sbb005821
82188
2-iodo-1-methyl-4-nitrobenzene
CC1=CC=C(C=C1I)[N+]([O-])=O

2-Iodo-4-nitrotoluene is used as a pharmaceutical intermediate, chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iodo-4-nitrotoluene is used as a pharmaceutical intermediate, chemical synthesis intermediate.

Solubility
Soluble in toluene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents. Light sensitive.
TRUSTED_SUSTAINABILITY
Quantity 5 g
Sensitivity Light sensitive
Solubility Information Soluble in toluene.
Formula Weight 263.03
Percent Purity 97%
Chemical Name or Material 2-Iodo-4-nitrotoluene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'