missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-3-hydroxypyridine, 97%, ACROS Organics™

Brand:  Acros Organics 436390050

Code : 29

Additional Details : CAS Number : 40263-57-8 Weight : 0.05500kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10697220

  • £127.20 / 5g


Estimated Shipment
17-07-2018
Add to basket


Specifications

Chemical Identifiers

3-Hydroxy-2-iodopyridine
96.0
≥96.0%
C5H4INO
2-Iodo-3-pyridinol
40263-57-8
100.0
97%
220.99

Specifications

3-Hydroxy-2-iodopyridine
96.0
26.08 to 28.25% C, 6.09 to 6.59% N
Glass bottle
≥96.0%
C5H4INO
White to tan
2-Iodo-3-pyridinol
40263-57-8
100.0
Authentic
Crystalline Powder
97%
220.99
5g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Iodo-3-hydroxypyridine, 97%

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS