missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Iodo-2-methylpropane, 95%, stab. with copper powder, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
2-Iodo-2-methylpropane is used as a chemical reagent in organic synthesis. Further, it serves as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Iodo-2-methylpropane is used as a chemical reagent in organic synthesis. Further, it serves as a pharmaceutical intermediate.
Solubility
Miscible with ethanol and ether.
Notes
Light sensitive. Incompatible with strong oxidizing agents and strong bases.
2-Iodo-2-methylpropane is used as a chemical reagent in organic synthesis. Further, it serves as a pharmaceutical intermediate.
Solubility
Miscible with ethanol and ether.
Notes
Light sensitive. Incompatible with strong oxidizing agents and strong bases.
Specifications
Melting Point | -38°C |
Density | 1.547 |
Boiling Point | 99°C to 100°C |
Flash Point | 7°C (44°F) |
Odor | Recognizable |
Refractive Index | 1.491 |
Quantity | 25 g |
UN Number | UN2391 |
Beilstein | 505950 |
Sensitivity | Light sensitive |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'