missing translation for 'onlineSavingsMsg'
Learn More

2-Indanylboronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Catalog Number p-7047630
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
608534-44-7
C15H21BO2
244.14
MFCD03788726
YRTKSYXHFWUPAZ-UHFFFAOYSA-N
2-indanylboronic acid pinacol ester, 2-2,3-dihydro-1h-inden-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-indanylboronicacidpinacolester, 2-indan-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
24208876
2-(2,3-dihydro-1H-inden-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
CC1(C)OB(OC1(C)C)C1CC2=CC=CC=C2C1
This item is not returnable. View return policy
608534-44-7
C15H21BO2
244.14
MFCD03788726
YRTKSYXHFWUPAZ-UHFFFAOYSA-N
2-indanylboronic acid pinacol ester, 2-2,3-dihydro-1h-inden-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-indanylboronicacidpinacolester, 2-indan-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
24208876
2-(2,3-dihydro-1H-inden-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
CC1(C)OB(OC1(C)C)C1CC2=CC=CC=C2C1

Boronic acids react with alcohols, with loss of water, to form boronic esters. With simple alcohols, the products are very susceptible to hydrolysis, but with 1,2- and 1,3-diols, the resulting cyclic boronates (1,3,2- dioxaborolanes and 1,3,2-dioxaborinanes) are stable enough to be isolated.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Boronic acids react with alcohols, with loss of water, to form boronic esters. With simple alcohols, the products are very susceptible to hydrolysis, but with 1,2- and 1,3-diols, the resulting cyclic boronates (1,3,2- dioxaborolanes and 1,3,2-dioxaborinanes) are stable enough to be isolated.

Solubility
Difficult to mix in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Refractive Index 1.504
Quantity 250 mg
Solubility Information Difficult to mix in water.
Formula Weight 244.14
Percent Purity 95%
Chemical Name or Material 2-Indanylboronic acid pinacol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'