missing translation for 'onlineSavingsMsg'
Learn More

2-Iminothiolane hydrochloride, Thermo Scientific Chemicals

Catalog Number p-7041757
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
4781-83-3
C4H8ClNS
137.625
MFCD00039013
ATGUDZODTABURZ-UHFFFAOYSA-N
2-iminothiolane hydrochloride, 2-iminothiolane hcl, thiolan-2-imine hydrochloride, dihydrothiophen-2 3h-imine hydrochloride, 2 3h-thiophenimine, dihydro-, hydrochloride, traut's reagent, c4h7ns.clh, traut eo 1/4 a, 2-thiolanimine hydrochloride, 2 3h-thiophenimine, hydrochloride
13166855
thiolan-2-imine;hydrochloride
C1CC(=N)SC1.Cl
This item is not returnable. View return policy
4781-83-3
C4H8ClNS
137.625
MFCD00039013
ATGUDZODTABURZ-UHFFFAOYSA-N
2-iminothiolane hydrochloride, 2-iminothiolane hcl, thiolan-2-imine hydrochloride, dihydrothiophen-2 3h-imine hydrochloride, 2 3h-thiophenimine, dihydro-, hydrochloride, traut's reagent, c4h7ns.clh, traut eo 1/4 a, 2-thiolanimine hydrochloride, 2 3h-thiophenimine, hydrochloride
13166855
thiolan-2-imine;hydrochloride
C1CC(=N)SC1.Cl

Protein modification and cross-linking reagent, cleavable by thiols

2-Iminothiolane hydrochloride is predominantly used as an amine reactive at pH 7-10 and is also an effective thiolation reagent for polysaccharides. It is also used as a RNA-protein crosslinking reagent as well as useful in cross-linking proteins..

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iminothiolane hydrochloride is predominantly used as an amine reactive at pH 7-10 and is also an effective thiolation reagent for polysaccharides. It is also used as a RNA-protein crosslinking reagent as well as useful in cross-linking proteins..

Solubility
Freely soluble in water

Notes
Protect from moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away fom strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 190°C to 195°C
Color White
Quantity 250 mg
Beilstein 3620079
Solubility Information Freely soluble in water
Formula Weight 137.63
Physical Form Powder
Chemical Name or Material 2-Iminothiolane hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'