Learn More
2-Iminopiperidine hydrochloride, Tocris Bioscience™
Selective iNOS inhibitor
Brand:  Tocris (Bio-Techne) 095110
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13458193
  • £85.05 / 10mg
Estimated Shipment
07-03-2019
Add to basket
Description
Description
An inhibitor of NO synthases, more potent than NG-methyl-L-arginine and selective for iNOS.
Specifications
Chemical Identifiers
16011-96-4
134.607
2-iminopiperidine hydrochloride, piperidin-2-imine hydrochloride, piperidine, 2-imino-, hydrochloride, 2-amino-3,4,5,6-tetrahydropyridine, piperidine, 2-imino-, monohydrochloride, 2-amino-3,4,5,6-tetrahydropyridine hydrochloride, piperidin-2-imine hydrochloride 1:1, 2-iminopiperidine hydro-chloride, 2-pyridinamine, 3,4,5,6-tetrahydro-, monohydrochloride, piperidin-2-ylideneamine hydrochloride
2,3,4,5-tetrahydropyridin-6-amine;hydrochloride
C5H11ClN2
ZHDTXTDHBRADLM-UHFFFAOYSA-N
85236
C1CCN=C(C1)N.Cl
Specifications
2-Iminopiperidine hydrochloride
16011-96-4
2-iminopiperidine hydrochloride, piperidin-2-imine hydrochloride, piperidine, 2-imino-, hydrochloride, 2-amino-3,4,5,6-tetrahydropyridine, piperidine, 2-imino-, monohydrochloride, 2-amino-3,4,5,6-tetrahydropyridine hydrochloride, piperidin-2-imine hydrochloride 1:1, 2-iminopiperidine hydro-chloride, 2-pyridinamine, 3,4,5,6-tetrahydro-, monohydrochloride, piperidin-2-ylideneamine hydrochloride
C1CCN=C(C1)N.Cl
134.607
134.61
10mg
C5H11ClN2
ZHDTXTDHBRADLM-UHFFFAOYSA-N
2,3,4,5-tetrahydropyridin-6-amine;hydrochloride
85236
>98%
Videos