missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxysaclofen, Tocris Bioscience™

Selective GABAB antagonist; more potent than Saclofen (Cat. No. 0246)

Brand:  Tocris (Bio-Techne) 024550

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13467503

  • £771.75 / 50mg

Estimated Shipment date
from Supplier 31-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective antagonist at GABAB receptors.
Specifications

Chemical Identifiers

117354-64-0
265.708
2-hydroxysaclofen, 2-oh-saclofen, 2-hydroxy-saclofen, 3-amino-2-4-chlorophenyl-2-hydroxypropane-1-sulfonic acid, 2-hydroxy-s---saclofen, 3-amino-2-4-chlorophenyl-2-hydroxypropylsulfonic acid, 3-amino-2-4-chlorophenyl-2-hydroxypropanesulfonic acid, beta-aminomethyl-4-chloro-beta-hydroxybenzeneethanesulfonic acid, benzeneethanesulfonic acid, beta-aminomethyl-4-chloro-beta-hydroxy, 2-hydroxysaclofen, solid
3-amino-2-(4-chlorophenyl)-2-hydroxypropane-1-sulfonic acid
C9H12ClNO4S
WBSMZVIMANOCNX-UHFFFAOYSA-N
1564
C1=CC(=CC=C1C(CN)(CS(=O)(=O)O)O)Cl

Specifications

50mg
117354-64-0
2-hydroxysaclofen, 2-oh-saclofen, 2-hydroxy-saclofen, 3-amino-2-4-chlorophenyl-2-hydroxypropane-1-sulfonic acid, 2-hydroxy-s---saclofen, 3-amino-2-4-chlorophenyl-2-hydroxypropylsulfonic acid, 3-amino-2-4-chlorophenyl-2-hydroxypropanesulfonic acid, beta-aminomethyl-4-chloro-beta-hydroxybenzeneethanesulfonic acid, benzeneethanesulfonic acid, beta-aminomethyl-4-chloro-beta-hydroxy, 2-hydroxysaclofen, solid
C1=CC(=CC=C1C(CN)(CS(=O)(=O)O)O)Cl
265.708
265.71
2-Hydroxysaclofen
C9H12ClNO4S
WBSMZVIMANOCNX-UHFFFAOYSA-N
3-amino-2-(4-chlorophenyl)-2-hydroxypropane-1-sulfonic acid
1564
>98%
Videos
Documents