missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxypyridine N-oxide, 98+%, Thermo Scientific Chemicals

Catalog Number p-7035972
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
13161-30-3
C5H5NO2
111.1
MFCD00006195
SNUSZUYTMHKCPM-UHFFFAOYSA-N
2-pyridinol-1-oxide, 2-hydroxypyridine 1-oxide, 2-hydroxypyridine n-oxide, 1-hydroxypyridin-2 1h-one, 2-hydroxypyridine-n-oxide, hydroxypyridinone, pyridinol, 1-oxide, 2-pyridinol 1-oxide, 1-hydroxy-2-pyridone, 1-hydroxy-1,2-dihydropyridin-2-one
69975
1-hydroxypyridin-2-one
C1=CC(=O)N(C=C1)O
This item is not returnable. View return policy
13161-30-3
C5H5NO2
111.1
MFCD00006195
SNUSZUYTMHKCPM-UHFFFAOYSA-N
2-pyridinol-1-oxide, 2-hydroxypyridine 1-oxide, 2-hydroxypyridine n-oxide, 1-hydroxypyridin-2 1h-one, 2-hydroxypyridine-n-oxide, hydroxypyridinone, pyridinol, 1-oxide, 2-pyridinol 1-oxide, 1-hydroxy-2-pyridone, 1-hydroxy-1,2-dihydropyridin-2-one
69975
1-hydroxypyridin-2-one
C1=CC(=O)N(C=C1)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 147°C to 152°C
Density 1.111
Flash Point 100°C (212°F)
Quantity 5 g
Beilstein 1422545
Formula Weight 111.1
Percent Purity ≥98%
Chemical Name or Material 2-Hydroxypyridine N-oxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'