missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxypyridine, 98%, Thermo Scientific Chemicals

Catalog Number p-7021966
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
142-08-5
C5H5NO
95.10
MFCD00006268
UBQKCCHYAOITMY-UHFFFAOYSA-N
2-hydroxypyridine, pyridin-2-ol, 2-pyridinol, 2-pyridone, pyridin-2 1h-one, 2 1h-pyridinone, 2-pyridinone, 2-oxopyridine, 2 1h-pyridone, alpha-pyridone
8871
CHEBI:16540
O=C1NC=CC=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
142-08-5
C5H5NO
95.10
MFCD00006268
UBQKCCHYAOITMY-UHFFFAOYSA-N
2-hydroxypyridine, pyridin-2-ol, 2-pyridinol, 2-pyridone, pyridin-2 1h-one, 2 1h-pyridinone, 2-pyridinone, 2-oxopyridine, 2 1h-pyridone, alpha-pyridone
8871
CHEBI:16540
O=C1NC=CC=C1

2-Hydroxypyridine is a bifunctional catalyst for a wide variety of acylation reactions, catalyst for generating β-oxopropyl carbonates from cyclic carbonates and alcohols and in the aminolysis of a polyglutamate. It is used in peptide synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxypyridine is a bifunctional catalyst for a wide variety of acylation reactions, catalyst for generating β-oxopropyl carbonates from cyclic carbonates and alcohols and in the aminolysis of a polyglutamate. It is used in peptide synthesis.

Solubility
Soluble in water 450 g/L @ 20°C and ethanol, slightly soluble in benzene and ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 102°C to 111°C
Density 1.39
Boiling Point 280°C to 281°C
Flash Point 210°C (410°F)
Odor Recognizable
Quantity 25 g
UN Number UN2811
Beilstein 105786
Solubility Information Soluble in water 450g/L @ 20°C and ethanol,slightly soluble in benzene and ether.
IUPAC Name 1H-pyridin-2-one
Formula Weight 95.1
Percent Purity 98%
Chemical Name or Material 2-Hydroxypyridine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'