missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxymethyl-18-crown-6, 97%, Thermo Scientific Chemicals

Catalog Number p-7059368
Click to view available options
Quantity:
0.25 g
1 g
This item is not returnable. View return policy
70069-04-4
C13H26O7
294.34
MFCD00188046
HFRGASADQCZXHH-UHFFFAOYNA-N
1,4,7,10,13,16-hexaoxacyclooctadecane-2-methanol, 2-hydroxymethyl-18-crown-6, 2-hydroxymethyl-18-crown 6-ether, 1,4,7,10,13,16-hexaoxacyclooctadecan-2-ylmethanol, hydroxy-18-crown-6, 18-crown-6-methanol, hydroxymethyl-18-crown-6, hfrgasadqczxhh-uhfffaoysa, 2-hydroxymethyl-18-crown, 2,5,8,11,14,17-hexaoxacyclooctadecyl methan-1-ol
3611393
1,4,7,10,13,16-hexaoxacyclooctadec-2-ylmethanol
OCC1COCCOCCOCCOCCOCCO1
This item is not returnable. View return policy
70069-04-4
C13H26O7
294.34
MFCD00188046
HFRGASADQCZXHH-UHFFFAOYNA-N
1,4,7,10,13,16-hexaoxacyclooctadecane-2-methanol, 2-hydroxymethyl-18-crown-6, 2-hydroxymethyl-18-crown 6-ether, 1,4,7,10,13,16-hexaoxacyclooctadecan-2-ylmethanol, hydroxy-18-crown-6, 18-crown-6-methanol, hydroxymethyl-18-crown-6, hfrgasadqczxhh-uhfffaoysa, 2-hydroxymethyl-18-crown, 2,5,8,11,14,17-hexaoxacyclooctadecyl methan-1-ol
3611393
1,4,7,10,13,16-hexaoxacyclooctadec-2-ylmethanol
OCC1COCCOCCOCCOCCOCCO1

2-Hydroxymethyl-18-crown-6 is used for chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.174 g/mL
Boiling Point 150°C (0.5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.479
Quantity 0.25 g
Beilstein 4313301
Solubility Information Soluble in water (1000g/L) (25°C).
Formula Weight 294.35
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Hydroxymethyl-18-crown-6
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'