missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxyethyl stearate, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
250 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 55°C to 60°C |
Color | White to Yellow |
Boiling Point | 149°C |
Infrared Spectrum | Authentic |
Packaging | Glass bottle |
Quantity | 100 g |
Solubility Information | Solubility in water: insoluble in h2o |
Formula Weight | 328.53 |
Grade | Technical |
Concentration or Composition (by Analyte or Components) | compound is a mixture of various chain-length esters and di(ethylene glycol) ester |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'