missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyethyl acrylate, 97%, stabilized, Thermo Scientific Chemicals

Catalog Number 10786232
Click to view available options
Quantity:
5 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
818-61-1, 150-76-5
C5H8O3
116.12
MFCD00002865
OMIGHNLMNHATMP-UHFFFAOYSA-N
2-hydroxyethyl acrylate, hydroxyethyl acrylate, 2-propenoic acid, 2-hydroxyethyl ester, ethylene glycol monoacrylate, bisomer 2hea, acrylic acid 2-hydroxyethyl ester, 2-acryloyloxy ethanol, ethylene glycol, acrylate, acrylic acid, 2-hydroxyethyl ester, 2-hydroxyethylacrylate
13165
2-hydroxyethyl prop-2-enoate
C=CC(=O)OCCO
This item is not returnable. View return policy
818-61-1, 150-76-5
C5H8O3
116.12
MFCD00002865
OMIGHNLMNHATMP-UHFFFAOYSA-N
2-hydroxyethyl acrylate, hydroxyethyl acrylate, 2-propenoic acid, 2-hydroxyethyl ester, ethylene glycol monoacrylate, bisomer 2hea, acrylic acid 2-hydroxyethyl ester, 2-acryloyloxy ethanol, ethylene glycol, acrylate, acrylic acid, 2-hydroxyethyl ester, 2-hydroxyethylacrylate
13165
2-hydroxyethyl prop-2-enoate
C=CC(=O)OCCO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Acidity 4mg KOH/g max.
CAS Min % 96.0
CAS Max % 100.0
Melting Point -60°C
Color Yellow to Brown
Density 1.1060g/mL
Boiling Point 210°C to 215°C
Flash Point 99°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4490 to 1.451
Quantity 5 mL
Specific Gravity 1.106
Solubility Information Solubility in water: soluble in water
Stabilizer 200 to 450ppm MEHQ
Viscosity 9 mPa.s (20°C)
Formula Weight 116.12
Percent Purity 97%
Physical Form Oily Liquid
Chemical Name or Material 2-Hydroxyethyl acrylate, Stabilized
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Hydroxyethyl acrylate, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P361-Take off immediately all contaminated clothing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'