missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxyacetophenone, 97+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
Description
2-Hydroxyacetophenone can be used to produce 2-hydroxy-1-phenyl-ethanone-(1)-oxime by heating. It is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon
Specifications
CAS | 582-24-1 |
Melting Point | 86°C to 88°C |
Molecular Formula | C8H8O2 |
MDL Number | MFCD00041829 |
Quantity | 5 g |
Sensitivity | Air and moisture sensitive |
Synonym | 2-hydroxyacetophenone, benzoylcarbinol, glycolophenone, phenacyl alcohol, alpha-hydroxyacetophenone, 2-hydroxy-1-phenylethan-1-one, ethanone, 2-hydroxy-1-phenyl, methanol, benzoyl, omega-hydroxyacetophenone, acetophenone, 2-hydroxy |
Solubility Information | Soluble in water,ether,ethanol,chloroform. |
InChI Key | ZWVHTXAYIKBMEE-UHFFFAOYSA-N |
SMILES | C1=CC=C(C=C1)C(=O)CO |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'