missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2-Hydroxy-p-naphthoquinone, 99%

Catalog Number p-146540
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
83-72-7
C10H6O3
174.16
MFCD00001678
WVCHIGAIXREVNS-UHFFFAOYSA-N
lawsone, 2-hydroxy-1,4-naphthoquinone, 2-hydroxynaphthalene-1,4-dione, 2-hydroxynaphthoquinone, henna, mehendi, hana, lawson, mendi, flower of paradise
6755
4-hydroxynaphthalene-1,2-dione
C1=CC=C2C(=C1)C(=CC(=O)C2=O)O
This item is not returnable. View return policy
83-72-7
C10H6O3
174.16
MFCD00001678
WVCHIGAIXREVNS-UHFFFAOYSA-N
lawsone, 2-hydroxy-1,4-naphthoquinone, 2-hydroxynaphthalene-1,4-dione, 2-hydroxynaphthoquinone, henna, mehendi, hana, lawson, mendi, flower of paradise
6755
4-hydroxynaphthalene-1,2-dione
C1=CC=C2C(=C1)C(=CC(=O)C2=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 192.0°C to 195.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 10 g
Beilstein 08, 300
Fieser 01,484
Merck Index 15, 5448
Solubility Information Solubility in water: 2g/L water (20°C). Other solubilities: soluble in benzene,carbon tetrachloride,aceton,,petroleum ether and sodium hydroxide
Formula Weight 174.16
Percent Purity 99%
Grade Pure
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-p-naphthoquinone, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'