missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-9-fluorenone, 97%, Thermo Scientific Chemicals

Catalog Number p-7052642
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
6949-73-1
C13H8O2
196.205
MFCD00001155
GXUBPHMYNSICJC-UHFFFAOYSA-N
2-hydroxy-9-fluorenone, 2-hydroxy-9h-fluoren-9-one, 2-hydroxy-fluoren-9-one, acmc-209o8z, 9h-fluoren-9-one,2-hydroxy, 2-hydroxy-9h-fluoren-9-one #, 9h-fluoren-9-one, 2-hydroxy
81386
2-hydroxyfluoren-9-one
C1=CC=C2C(=C1)C3=C(C2=O)C=C(C=C3)O
This item is not returnable. View return policy
6949-73-1
C13H8O2
196.205
MFCD00001155
GXUBPHMYNSICJC-UHFFFAOYSA-N
2-hydroxy-9-fluorenone, 2-hydroxy-9h-fluoren-9-one, 2-hydroxy-fluoren-9-one, acmc-209o8z, 9h-fluoren-9-one,2-hydroxy, 2-hydroxy-9h-fluoren-9-one #, 9h-fluoren-9-one, 2-hydroxy
81386
2-hydroxyfluoren-9-one
C1=CC=C2C(=C1)C3=C(C2=O)C=C(C=C3)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 204°C to 206°C
Color Yellow to Orange
Quantity 250 mg
Formula Weight 196.21
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2-Hydroxy-9-fluorenone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'