missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Hydroxy-6-(trifluoromethyl)nicotinic acid, 95%

Brand:  Alfa Aesar™ H64060.03

Code : NEW

Additional Details : CAS Number : 191595-63-8 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15441089

  • £86.07 / 1g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

2-Hydroxy-6-(trifluoromethyl)nicotinic acid
207.11
2-Hydroxy-6-(trifluoromethyl)nicotinic acid
191595-63-8
MFCD01862657

Specifications

2-Hydroxy-6-(trifluoromethyl)nicotinic acid
191595-63-8
95%
MFCD01862657
1g
Solid
207.11
2-Hydroxy-6-(trifluoromethyl)nicotinic acid
Videos
SDS