missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-5-(trifluoromethyl)pyridine, 97%, Thermo Scientific™

Catalog Number p-3758214
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
33252-63-0
C6H4F3NO
163.10
MFCD00042315
BYRJSCNPUHYZQE-UHFFFAOYSA-N
2-hydroxy-5-trifluoromethylpyridine, 5-trifluoromethyl pyridin-2-ol, 2-hydroxy-5-trifluoromethyl pyridine, 5-trifluoromethyl-2-pyridinol, 5-trifluoromethyl-pyridin-2-ol, 5-trifluoromethyl-2-pyridone, unii-1ccp81h07b, 5-trifluoromethyl pyridin-2 1h-one, 5-trifluoromethyl-1,2-dihydropyridin-2-one, 5-trifloromethyl-2-1h-pyridinone
147443
5-(trifluoromethyl)-1H-pyridin-2-one
FC(F)(F)C1=CNC(=O)C=C1
This item is not returnable. View return policy
33252-63-0
C6H4F3NO
163.10
MFCD00042315
BYRJSCNPUHYZQE-UHFFFAOYSA-N
2-hydroxy-5-trifluoromethylpyridine, 5-trifluoromethyl pyridin-2-ol, 2-hydroxy-5-trifluoromethyl pyridine, 5-trifluoromethyl-2-pyridinol, 5-trifluoromethyl-pyridin-2-ol, 5-trifluoromethyl-2-pyridone, unii-1ccp81h07b, 5-trifluoromethyl pyridin-2 1h-one, 5-trifluoromethyl-1,2-dihydropyridin-2-one, 5-trifloromethyl-2-1h-pyridinone
147443
5-(trifluoromethyl)-1H-pyridin-2-one
FC(F)(F)C1=CNC(=O)C=C1
CAS Min % 96.0
CAS Max % 100.0
Melting Point 145.0°C to 149.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 163.1
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-5-(trifluoromethyl)pyridine
Show More Show Less
compliance-icons
Product Identifier
  • 2-Hydroxy-5-(trifluoromethyl)pyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'