missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-5-methoxybenzaldehyde, 98%, ACROS Organics™

Brand:  Acros Organics 164160100

Code : 29

Additional Details : CAS Number : 672-13-9 Weight : 0.06000kg

Quantity 10g
Packaging Glass bottle

 View more versions of this product

Product Code. 10617142

  • £53.90 / 10g


EU Stock 2
Add to basket

Specifications

Chemical Identifiers

2-Hydroxy-5-methoxybenzaldehyde
97.5
≥97.5%
C8H8O3
152.15
5-Methoxysalicylaldehyde
672-13-9
100.0
98%
HOC6H3(OCH3)CHO
MFCD00003332

Specifications

2-Hydroxy-5-methoxybenzaldehyde
Clear yellow
672-13-9
100.0
Glass bottle
≥97.5%
1.5770 to 1.5790 (20°C, 589nm)
HOC6H3(OCH3)CHO
MFCD00003332
5-Methoxysalicylaldehyde
1.21g/mL
10g
97.5
Authentic
Liquid
98%
C8H8O3
152.15
08, 244
Videos
SDS