missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-methylbenzaldehyde, 97%, ACROS Organics™

Catalog Number p-4228719
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
698-27-1
C8H8O2
136.15
MFCD00799550
JODRRPJMQDFCBJ-UHFFFAOYSA-N
4-methylsalicylaldehyde, 2,4-cresotaldehyde, benzaldehyde, 2-hydroxy-4-methyl, 4-methyl-2-hydroxybenzaldehyde, 2-hydroxy-4-methyl-benzaldehyde, 2-formyl-5-methylphenol, 4-methylsalicyclic aldehyde, unii-9n0s26g61j, 4-methyl salicylaldehyde, homosalicylaldehyde, m
61200
2-hydroxy-4-methylbenzaldehyde
CC1=CC=C(C=O)C(O)=C1
This item is not returnable. View return policy
698-27-1
C8H8O2
136.15
MFCD00799550
JODRRPJMQDFCBJ-UHFFFAOYSA-N
4-methylsalicylaldehyde, 2,4-cresotaldehyde, benzaldehyde, 2-hydroxy-4-methyl, 4-methyl-2-hydroxybenzaldehyde, 2-hydroxy-4-methyl-benzaldehyde, 2-formyl-5-methylphenol, 4-methylsalicyclic aldehyde, unii-9n0s26g61j, 4-methyl salicylaldehyde, homosalicylaldehyde, m
61200
2-hydroxy-4-methylbenzaldehyde
CC1=CC=C(C=O)C(O)=C1
CAS Min % 96.0
CAS Max % 100.0
Melting Point 59°C to 63°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 25 g
Formula Weight 136.15
Percent Purity ≥96.0%
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-4-methylbenzaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 2-Hydroxy-4-methylbenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'