missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-3-methylbenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog Number p-3759058
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
824-42-0
C8H8O2
136.15
MFCD00040798
IPPQNXSAJZOTJZ-UHFFFAOYSA-N
3-methylsalicylaldehyde, 2,3-cresotaldehyde, 2-hydroxy-3-methyl-benzaldehyde, 3-methylsalicylic aldehyde, 3-methyl-2-hydroxybenzaldehyde, benzaldehyde, hydroxy-3-methyl, benzaldehyde, 2-hydroxy-3-methyl, zlchem 747, pubchem14264, 2-formyl-6-methylphenol
522777
CHEBI:20110
2-hydroxy-3-methylbenzaldehyde
CC1=CC=CC(=C1O)C=O
This item is not returnable. View return policy
824-42-0
C8H8O2
136.15
MFCD00040798
IPPQNXSAJZOTJZ-UHFFFAOYSA-N
3-methylsalicylaldehyde, 2,3-cresotaldehyde, 2-hydroxy-3-methyl-benzaldehyde, 3-methylsalicylic aldehyde, 3-methyl-2-hydroxybenzaldehyde, benzaldehyde, hydroxy-3-methyl, benzaldehyde, 2-hydroxy-3-methyl, zlchem 747, pubchem14264, 2-formyl-6-methylphenol
522777
CHEBI:20110
2-hydroxy-3-methylbenzaldehyde
CC1=CC=CC(=C1O)C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Orange to Yellow
Density 1.1340g/mL
Boiling Point 203°C to 205°C (730.0 mmHg)
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H3(OH)CHO
Quantity 1 g
Specific Gravity 1.134
Formula Weight 136.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Hydroxy-3-methylbenzaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 2-Hydroxy-3-methylbenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'