missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydrazino-4-(trifluoromethyl)pyrimidine, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
2-Hydrazino-4-(trifluoromethyl)pyrimidine may be used as a derivatization reagent for the simultaneous determination of testosterone (T) and dihydrotestosterone (DHT) in murine tissue and serum. Synthesis of organohydrazide chelate complexes of rhenium and technetium. Preparation of 1-(4-methoxyphenyl)ethanone [(4-trifluoromethyl)pyrimidine-2-yl]hydrazone (NAS-75).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Hydrazino-4-(trifluoromethyl)pyrimidine may be used as a derivatization reagent for the simultaneous determination of testosterone (T) and dihydrotestosterone (DHT) in murine tissue and serum. Synthesis of organohydrazide chelate complexes of rhenium and technetium. Preparation of 1-(4-methoxyphenyl)ethanone [(4-trifluoromethyl)pyrimidine-2-yl]hydrazone (NAS-75).
Solubility
Soluble in aqueous acid.
Notes
Store in cool, dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
2-Hydrazino-4-(trifluoromethyl)pyrimidine may be used as a derivatization reagent for the simultaneous determination of testosterone (T) and dihydrotestosterone (DHT) in murine tissue and serum. Synthesis of organohydrazide chelate complexes of rhenium and technetium. Preparation of 1-(4-methoxyphenyl)ethanone [(4-trifluoromethyl)pyrimidine-2-yl]hydrazone (NAS-75).
Solubility
Soluble in aqueous acid.
Notes
Store in cool, dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
Specifications
Melting Point | 99°C to 101°C |
Quantity | 1 g |
UN Number | UN2811 |
Solubility Information | Soluble in aqueous acid. |
Formula Weight | 178.12 |
Percent Purity | 97% |
Chemical Name or Material | 2-Hydrazino-4-(trifluoromethyl)pyrimidine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'