missing translation for 'onlineSavingsMsg'
Learn More

2-Hexanone, 98%, Acros Organics

£65.70 - £882.50

Chemical Identifiers

CAS 591-78-6
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00009482
InChI Key QQZOPKMRPOGIEB-UHFFFAOYSA-N
Synonym 2-hexanone, hexanone, 2-oxohexane, propylacetone, n-butyl methyl ketone, butyl methyl ketone, methyl butyl ketone, methyl n-butyl ketone, hexanone-2, ketone, butyl methyl
PubChem CID 11583
IUPAC Name hexan-2-one
SMILES CCCCC(=O)C
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10386830 SDS View Documents Acros Organics
146881000
100mL Glass bottle £65.70
100mL View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 03-02-2020
Please to purchase this item. Need a web account? Register with us today!
10021590 SDS View Documents Acros Organics
146885000
500mL Glass bottle £225.80
500mL View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 31-01-2020
Please to purchase this item. Need a web account? Register with us today!
10207593 SDS View Documents Acros Organics
146880025
2.5L Glass bottle £882.50
2.50 L View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

591-78-6
100.16
QQZOPKMRPOGIEB-UHFFFAOYSA-N
11583
CCCCC(=O)C
C6H12O
MFCD00009482
2-hexanone, hexanone, 2-oxohexane, propylacetone, n-butyl methyl ketone, butyl methyl ketone, methyl butyl ketone, methyl n-butyl ketone, hexanone-2, ketone, butyl methyl
hexan-2-one

Specifications

2-Hexanone
23°C
Glass bottle
0.81
Colorless to Yellow
100mL
591-78-6
100.0
C6H12O
MFCD00009482
15, 6107
Solubility in water: 20g/l (20°C). Other solubilities: soluble in alcohol and ether
CCCCC(=O)C
100.16
100.16
98%
0.8100g/mL
Authentic
1.3995 to 1.4015
127°C
-57°C
98%
97.5
98%
CH3(CH2)3COCH3
01, 689
2-hexanone, hexanone, 2-oxohexane, propylacetone, n-butyl methyl ketone, butyl methyl ketone, methyl butyl ketone, methyl n-butyl ketone, hexanone-2, ketone, butyl methyl
QQZOPKMRPOGIEB-UHFFFAOYSA-N
hexan-2-one
11583
Liquid
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Hexanone

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 3
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H372-Causes damage to organs.
  • H361f-Suspected of damaging fertility.
  • H336-May cause drowsiness or dizziness.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.