missing translation for 'onlineSavingsMsg'
Learn More

2-Hexanol, 99%, ACROS Organics™

Brand:  Acros Organics 156290500

Code : 29

Additional Details : CAS Number : 626-93-7 Weight : 0.05000kg Transport : UN number : 2282 Chem class : 3 Pack group : III


Packaging Glass bottle
Quantity 50g

 View more versions of this product

Product Code. 10499242

  • £52.30 / 50g


EU Stock 2
Add to basket


Specifications

Chemical Identifiers

2-Hexanol
98.5
99%
C6H14O
102.18
626-93-7
100.0
99%
CH3(CH2)3CH(OH)CH3
MFCD00004585

Specifications

2-Hexanol
139.0°C
− 23.0°C
626-93-7
100.0
Authentic
Liquid
99%
1.4125 to 1.4155
102.18
Solubility in water: insoluble.
0.81
41°C
Clear colorless
50g
98.5
0.8100g/mL
Glass bottle
99%
C6H14O
CH3(CH2)3CH(OH)CH3
MFCD00004585
01, 408
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Hexanol

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS