missing translation for 'onlineSavingsMsg'
Learn More

2-Heptyne, 97%, Thermo Scientific™

Catalog Number p-7077563
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
1119-65-9
C7H12
96.17
AMSFEMSYKQQCHL-UHFFFAOYSA-N
2-heptyne, 1-methyl-2-butylacetylene, butylmethylacetylene, n-butylmethylacetylene, acmc-1brzr, n-c4h9c.$.cch3, 2-heptyne,
14245
hept-2-yne
CCCCC#CC
This item is not returnable. View return policy
1119-65-9
C7H12
96.17
AMSFEMSYKQQCHL-UHFFFAOYSA-N
2-heptyne, 1-methyl-2-butylacetylene, butylmethylacetylene, n-butylmethylacetylene, acmc-1brzr, n-c4h9c.$.cch3, 2-heptyne,
14245
hept-2-yne
CCCCC#CC
Density 0.7590g/mL
Boiling Point 110.0°C to 111.0°C (743.0 mmHg)
Flash Point 11°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Linear Formula CH3(CH2)3C≡CCH3
Quantity 5 g
Specific Gravity 0.759
Formula Weight 96.17
Percent Purity 97%
Chemical Name or Material 2-Heptyne
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Heptyne
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'