missing translation for 'onlineSavingsMsg'
Learn More

2-Heptanone, 98%, ACROS Organics™

£10.50 - £274.30

Chemical Identifiers

CAS 110-43-0
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.19
MDL Number MFCD00009513
InChI Key CATSNJVOTSVZJV-UHFFFAOYSA-N
Synonym 2-heptanone, methyl amyl ketone, butylacetone, methyl pentyl ketone, methyl n-amyl ketone, amyl methyl ketone, n-amyl methyl ketone, n-pentyl methyl ketone, heptanone, pentyl methyl ketone
PubChem CID 8051
ChEBI CHEBI:5672
IUPAC Name heptan-2-one
SMILES CCCCCC(=O)C
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10051750 SDS View Documents Acros Organics
154001000
100mL Glass bottle £10.50
100mL View alternative products
Save Up to 
»
EU Stock 7 Please to purchase this item. Need a web account? Register with us today!
10051760 SDS View Documents Acros Organics
154002500
250mL Glass bottle £20.20
250mL View alternative products
Save Up to 
»
EU Stock 3 Please to purchase this item. Need a web account? Register with us today!
10307380 SDS View Documents Acros Organics
154000010
1L Glass bottle £55.40
1 L View alternative products
Save Up to 
»
EU Stock 21 Please to purchase this item. Need a web account? Register with us today!
10237213 SDS View Documents Acros Organics
154000051
5L Plastic drum £274.30
5 L View alternative products
Save Up to 
»
EU Stock 3 Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

110-43-0
114.19
CATSNJVOTSVZJV-UHFFFAOYSA-N
8051
heptan-2-one
C7H14O
MFCD00009513
2-heptanone, methyl amyl ketone, butylacetone, methyl pentyl ketone, methyl n-amyl ketone, amyl methyl ketone, n-amyl methyl ketone, n-pentyl methyl ketone, heptanone, pentyl methyl ketone
CHEBI:5672
CCCCCC(=O)C

Specifications

2-Heptanone
39°C
Glass bottle
0.82
Colorless
100mL
97.5
98%
CH3(CH2)4COCH3
01, 699
2-heptanone, methyl amyl ketone, butylacetone, methyl pentyl ketone, methyl n-amyl ketone, amyl methyl ketone, n-amyl methyl ketone, n-pentyl methyl ketone, heptanone, pentyl methyl ketone
CATSNJVOTSVZJV-UHFFFAOYSA-N
heptan-2-one
8051
114.19
98%
0.8200g/mL
Authentic
1.4075 to 1.4095
149.0°C to 150.0°C
-35.0°C
110-43-0
100.0
C7H14O
MFCD00009513
15, 4698
Solubility in water: 4.3g/L (20°C). Other solubilities: soluble in alcohol and ether
CCCCCC(=O)C
114.19
CHEBI:5672
Liquid
Pure
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Heptanone

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3

Hazard Statement
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.