missing translation for 'onlineSavingsMsg'
Learn More

2-Heptanol, 99+%, Thermo Scientific Chemicals

Catalog Number p-140166
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy
543-49-7
C7H16O
116.20
MFCD00004587
CETWDUZRCINIHU-UHFFFAOYNA-N
2-heptanol, s-heptyl alcohol, 2-heptyl alcohol, 2-hydroxyheptane, amyl methyl carbinol, heptanol-2, methyl amyl carbinol, 1-methylhexanol, heptyl alcohol, sec, 2-heptanol natural
10976
heptan-2-ol
CCCCCC(C)O
This item is not returnable. View return policy
543-49-7
C7H16O
116.20
MFCD00004587
CETWDUZRCINIHU-UHFFFAOYNA-N
2-heptanol, s-heptyl alcohol, 2-heptyl alcohol, 2-hydroxyheptane, amyl methyl carbinol, heptanol-2, methyl amyl carbinol, 1-methylhexanol, heptyl alcohol, sec, 2-heptanol natural
10976
heptan-2-ol
CCCCCC(C)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Color Colorless
Density 0.8175g/mL
Boiling Point 160°C to 162°C
Flash Point 59°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)4CH(OH)CH3
Refractive Index 1.4200 to 1.422
Quantity 50 mL
Beilstein 01, 415
Merck Index 15, 4697
Specific Gravity 0.8175
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,ether,benzene,miscible with most common organic liquids
Formula Weight 116.2
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 2-Heptanol, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Heptanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H312-Harmful in contact with skin.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'