2-Furylmethyl isothiocyanate, ≥95%, Maybridge


Overview
Brand: Maybridge

Manufacturer Part Number: SB01625DA

1GR 2-Furylmethyl isothiocyanate, 95%

UNSPSC: 12350000

Code: 31

Additional Details:
Additional Details: CAS Number: 4650-60-6 Weight: 0.05100kg Transport: UN number : 2810 Chem class : 1 Pack group : III


Packaging Amber glass bottle
Quantity 1g

Product Code. 10364392

Quantity Price
1 £ 8.0 / 1g
Estimated Shipment date
from Supplier 27-03-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 2-Furylmethyl isothiocyanate
CAS 4650-60-6
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%+
Molecular Formula C6H5NOS
Formula Weight 139.18
MDL Number MFCD00014112

Specification

Formula Weight 139.18
Quantity 1g
Assay 95%
CAS Min % 95.0
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 4650-60-6
Assay Percent Range 95%+
MDL Number MFCD00014112
Packaging Amber glass bottle
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Molecular Formula C6H5NOS
Health Hazard 1 GHS Signal Word: Warning
Density 1.3900g/mL
Chemical Name or Material 2-Furylmethyl isothiocyanate
Documentation

Technical documents

Safety and Handling

GHS Signal Word: Warning

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.