missing translation for 'onlineSavingsMsg'
Learn More

2-Furoyl chloride, 98+%, Thermo Scientific Chemicals

Catalog Number 10478910
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
527-69-5
C5H3ClO2
130.53
MFCD00003228
OFTKFKYVSBNYEC-UHFFFAOYSA-N
2-furoyl chloride, furoyl chloride, 2-furancarbonyl chloride, furancarbonyl chloride, 2-furoylchloride, unii-2us0dxi75s, 2us0dxi75s, 2-furancarboxylic acid chloride, furan-2-carboxylic acid chloride, furanoyl chloride
68242
furan-2-carbonyl chloride
C1=COC(=C1)C(=O)Cl
This item is not returnable. View return policy
527-69-5
C5H3ClO2
130.53
MFCD00003228
OFTKFKYVSBNYEC-UHFFFAOYSA-N
2-furoyl chloride, furoyl chloride, 2-furancarbonyl chloride, furancarbonyl chloride, 2-furoylchloride, unii-2us0dxi75s, 2us0dxi75s, 2-furancarboxylic acid chloride, furan-2-carboxylic acid chloride, furanoyl chloride
68242
furan-2-carbonyl chloride
C1=COC(=C1)C(=O)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Melting Point -2°C
Color Yellow to Brown
Density 1.3200g/mL
Boiling Point 173°C to 174°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.5300 to 1.532
Quantity 100 mL
Beilstein 18, 276
Merck Index 15, 4339
Specific Gravity 1.32
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in ether,toluene,methylene chloride,,dichloroethane,chlorobenzene,ethyl acetate,thf,and chloroform,decomposes in alcohol
Formula Weight 130.53
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 2-Furoyl chloride, 98+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Furoyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'