missing translation for 'onlineSavingsMsg'
Learn More

2-Furoic acid, 98%, Acros Organics

£9.10 - £358.70

Chemical Identifiers

CAS 88-14-2
Molecular Formula C5H4O3
Molecular Weight (g/mol) 112.08
MDL Number MFCD00003238
InChI Key SMNDYUVBFMFKNZ-UHFFFAOYSA-N
Synonym 2-furoic acid, 2-furancarboxylic acid, pyromucic acid, 2-carboxyfuran, furoic acid, furancarboxylic acid, alpha-furoic acid, alpha-furancarboxylic acid, 2-furanoic acid, kyselina 2-furoova
PubChem CID 6919
ChEBI CHEBI:30845
IUPAC Name furan-2-carboxylic acid
SMILES C1=COC(=C1)C(=O)O
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10070920 SDS View Documents Acros Organics
119820050
5g Glass bottle £9.10
5g View alternative products
Save Up to 
»
EU Stock 2 Please to purchase this item. Need a web account? Register with us today!
10497300 SDS View Documents Acros Organics
119821000
100g Plastic bottle £17.80
100g View alternative products
Save Up to 
»
EU Stock 1 Please to purchase this item. Need a web account? Register with us today!
10754231 SDS View Documents Acros Organics
119825000
500g Plastic bottle £77.80
500g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 06-03-2020
Please to purchase this item. Need a web account? Register with us today!
10309323 SDS View Documents Acros Organics
119820025
2.5kg Plastic bottle £358.70
2.50kg View alternative products
Save Up to 
»
EU Stock 1 Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

88-14-2
112.08
SMNDYUVBFMFKNZ-UHFFFAOYSA-N
6919
furan-2-carboxylic acid
C5H4O3
MFCD00003238
2-furoic acid, 2-furancarboxylic acid, pyromucic acid, 2-carboxyfuran, furoic acid, furancarboxylic acid, alpha-furoic acid, alpha-furancarboxylic acid, 2-furanoic acid, kyselina 2-furoova
CHEBI:30845
C1=COC(=C1)C(=O)O

Specifications

2-Furoic acid
Authentic
230.0°C to 232.0°C
129.0°C to 133.0°C
98%
97.5
98%
MFCD00003238
15, 4336
Solubility in water: 36g/L (20°C). Other solubilities: soluble in alcohol and ether,insoluble in paraffin hydrocarbons
C1=COC(=C1)C(=O)O
112.08
CHEBI:30845
Fine Crystalline Powder
137°C
Glass bottle
Beige to White
5g
88-14-2
100.0
C5H4O3
18, 272
2-furoic acid, 2-furancarboxylic acid, pyromucic acid, 2-carboxyfuran, furoic acid, furancarboxylic acid, alpha-furoic acid, alpha-furancarboxylic acid, 2-furanoic acid, kyselina 2-furoova
SMNDYUVBFMFKNZ-UHFFFAOYSA-N
furan-2-carboxylic acid
6919
112.08
98%
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Furoic acid

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.