missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2-Furanylmethylene)malononitrile, 98+%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Melting Point | 70°C to 74°C |
Assay Percent Range | 98% min. (GC) |
Packaging | Glass bottle |
Quantity | 25 g |
IUPAC Name | 2-[(furan-2-yl)methylidene]propanedinitrile |
Formula Weight | 114.13 |
Percent Purity | 98+% |
Chemical Name or Material | (2-Furanylmethylene)malononitrile |
Safety and Handling
Product Identifier
- (2-Furanylmethylene)malononitrile
- Warning
- Acute toxicity Category 4
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'