missing translation for 'onlineSavingsMsg'
Learn More

2-Furaldehyde, ACS, 98% min, Thermo Scientific Chemicals

Catalog Number p-7049925
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy
98-01-1
C5H4O2
96.09
MFCD00003229
HYBBIBNJHNGZAN-UHFFFAOYSA-N
furfural, 2-furaldehyde, 2-furancarboxaldehyde, furaldehyde, 2-formylfuran, furfuraldehyde, fural, 2-furanaldehyde, 2-furancarbonal, 2-furfural
7362
CHEBI:34768
furan-2-carbaldehyde
O=CC1=CC=CO1
This item is not returnable. View return policy
98-01-1
C5H4O2
96.09
MFCD00003229
HYBBIBNJHNGZAN-UHFFFAOYSA-N
furfural, 2-furaldehyde, 2-furancarboxaldehyde, furaldehyde, 2-formylfuran, furfuraldehyde, fural, 2-furanaldehyde, 2-furancarbonal, 2-furfural
7362
CHEBI:34768
furan-2-carbaldehyde
O=CC1=CC=CO1

It is primarily used in the production of furan derivatives such as furan and furfuryl alcohol. Another major application of furfural is as extraction solvent in refineries. It is also used to accelerate vulcanization, as an ingredient of phenolic resins (Durite) and as a flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -38°C
Color Yellow to Brown
Density 1.159 g/mL
Boiling Point 162°C
Flash Point 60°C (140°F)
Odor Almond-like
Refractive Index 1.5261
Quantity 250 g
UN Number UN1199
Beilstein 105755
Sensitivity Air Sensitive
Merck Index 14,4304
Solubility Information It is soluble in water.
Formula Weight 96.09
Percent Purity ≤98%
Grade ACS
Physical Form Liquid
Chemical Name or Material 2-Furaldehyde
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'