missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoropyridine, 99%, Thermo Scientific Chemicals

Catalog Number p-7081593
15.93 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
10 g
25 g
100 g
250 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
372-48-5
C5H4FN
97.092
MFCD00006224
MTAODLNXWYIKSO-UHFFFAOYSA-N
pyridine, 2-fluoro, o-fluoropyridine, 2-fluoro-pyridine, pyridine, fluoro, ccris 1725, 2-fluoranylpyridine, fluoropyridine, 2fluoropyridine, 2-fluorpyridin, 2-fluoro-pyridinium
9746
2-fluoropyridine
C1=CC=NC(=C1)F
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
372-48-5
C5H4FN
97.092
MFCD00006224
MTAODLNXWYIKSO-UHFFFAOYSA-N
pyridine, 2-fluoro, o-fluoropyridine, 2-fluoro-pyridine, pyridine, fluoro, ccris 1725, 2-fluoranylpyridine, fluoropyridine, 2fluoropyridine, 2-fluorpyridin, 2-fluoro-pyridinium
9746
2-fluoropyridine
C1=CC=NC(=C1)F

2-Fluoropyridine is a useful precursor of 2,3-disubstituted pyridines which is made by the possibility of metallation at C-3, together with the ready nucleophilic displacement of the activated fluorine. It can also react with thiophene to get 2-thiophen-2-yl-pyridine. It is used in the synthesis of aminopyridines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoropyridine is a useful precursor of 2,3-disubstituted pyridines which is made by the possibility of metallation at C-3, together with the ready nucleophilic displacement of the activated fluorine. It can also react with thiophene to get 2-thiophen-2-yl-pyridine. It is used in the synthesis of aminopyridines.

Solubility
Soluble in water.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 1.129
Boiling Point 124°C to 126°C
Flash Point 24°C (75°F)
Refractive Index 1.467
Quantity 10 g
UN Number UN1993
Beilstein 1515
Solubility Information Soluble in water.
Formula Weight 97.09
Percent Purity 99%
Chemical Name or Material 2-Fluoropyridine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'