missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoropyridine, 98%, Thermo Scientific Chemicals

Catalog Number 10454785
Click to view available options
Quantity:
5 mL
50 mL
250 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
372-48-5
C5H4FN
97.09
MFCD00006224
MTAODLNXWYIKSO-UHFFFAOYSA-N
pyridine, 2-fluoro, o-fluoropyridine, 2-fluoro-pyridine, pyridine, fluoro, ccris 1725, 2-fluoranylpyridine, fluoropyridine, 2fluoropyridine, 2-fluorpyridin, 2-fluoro-pyridinium
9746
2-fluoropyridine
C1=CC=NC(=C1)F
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
372-48-5
C5H4FN
97.09
MFCD00006224
MTAODLNXWYIKSO-UHFFFAOYSA-N
pyridine, 2-fluoro, o-fluoropyridine, 2-fluoro-pyridine, pyridine, fluoro, ccris 1725, 2-fluoranylpyridine, fluoropyridine, 2fluoropyridine, 2-fluorpyridin, 2-fluoro-pyridinium
9746
2-fluoropyridine
C1=CC=NC(=C1)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1200g/mL
Boiling Point 126.0°C (753.0 mmHg)
Flash Point 24°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (C5H4N)F
Refractive Index 1.4670 to 1.4690
Quantity 250 mL
Beilstein 20, I, 80
Specific Gravity 1.12
Formula Weight 97.09
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Fluoropyridine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Fluoropyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'