missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoropyridine-4-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Catalog Number p-7029687
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
458532-86-0
C11H15BFNO2
223.05
MFCD06798253
PCLMNCBIXQQRMB-UHFFFAOYSA-N
2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-fluoropyridine-4-boronic acid pinacol ester, 2-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-fluoropyridin-4-boronic acid pinacol ester, 2-fluoropyridin-4-ylboronic acid pinacol ester, pyridine, 2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, abbypharma ap-11-5396, 2-fluoro-4-pyridineboronic acid pinacol ester, 2-fluoropyridin-4-yl boronic acid pinacol ester, 2-2-fluoro 4-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
12060164
2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
CC1(C)OB(OC1(C)C)C1=CC(F)=NC=C1
This item is not returnable. View return policy
458532-86-0
C11H15BFNO2
223.05
MFCD06798253
PCLMNCBIXQQRMB-UHFFFAOYSA-N
2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-fluoropyridine-4-boronic acid pinacol ester, 2-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-fluoropyridin-4-boronic acid pinacol ester, 2-fluoropyridin-4-ylboronic acid pinacol ester, pyridine, 2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, abbypharma ap-11-5396, 2-fluoro-4-pyridineboronic acid pinacol ester, 2-fluoropyridin-4-yl boronic acid pinacol ester, 2-2-fluoro 4-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
12060164
2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
CC1(C)OB(OC1(C)C)C1=CC(F)=NC=C1

Used in pharmaceuticals, agrochemicals, and high-tech materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in pharmaceuticals, agrochemicals, and high-tech materials.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 48°C to 52°C
Quantity 1 g
Formula Weight 223.06
Percent Purity 95%
Chemical Name or Material 2-Fluoropyridine-4-boronic acid pinacol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'