missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoropyridine-3-carboxylic acid, 97%, Thermo Scientific™

Catalog Number 10862071
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
393-55-5
C6H4FNO2
141.1
LLLVHTWJGWNRBD-UHFFFAOYSA-N
2-fluoronicotinic acid, 2-fluoro-3-pyridinecarboxylic acid, 2-fluoro-nicotinic acid, 2-fluoro-3-carboxypyridine, 2-fluoronicotinicacid, 3-pyridinecarboxylic acid, 2-fluoro, 2-fluoro-3-pyridinecarboxylic acid 2-fluoronicotinic acid, fluoronicotinic acid, pubchem1120, 2-fluoro nicotinic acid
96089
2-fluoropyridine-3-carboxylic acid
C1=CC(=C(N=C1)F)C(=O)O
This item is not returnable. View return policy
393-55-5
C6H4FNO2
141.1
LLLVHTWJGWNRBD-UHFFFAOYSA-N
2-fluoronicotinic acid, 2-fluoro-3-pyridinecarboxylic acid, 2-fluoro-nicotinic acid, 2-fluoro-3-carboxypyridine, 2-fluoronicotinicacid, 3-pyridinecarboxylic acid, 2-fluoro, 2-fluoro-3-pyridinecarboxylic acid 2-fluoronicotinic acid, fluoronicotinic acid, pubchem1120, 2-fluoro nicotinic acid
96089
2-fluoropyridine-3-carboxylic acid
C1=CC(=C(N=C1)F)C(=O)O
CAS Min % 96.0
CAS Max % 100.0
Melting Point 150.8°C to 151.4°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 141.1
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 2-Fluoropyridine-3-carboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Fluoropyridine-3-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'