missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorophenethyl bromide, 97%, Thermo Scientific Chemicals

Catalog Number p-4228686
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
91319-54-9
C8H8BrF
203.05
FQGDFWIQBCQXPS-UHFFFAOYSA-N
2-fluorophenethyl bromide, 1-2-bromoethyl-2-fluorobenzene, 1-2-bromo-ethyl-2-fluoro-benzene, benzene, 1-2-bromoethyl-2-fluoro, 2-2-bromoethyl-1-fluorobenzene, 1-2-bromoethyl-2-fluoro-benzene, 2-fluorophenyl-ethylbromide, acmc-209wk4, 2-fluorophenyl ethyl bromide, 2-2-fluorophenyl-ethylbromide
2756964
1-(2-bromoethyl)-2-fluorobenzene
C1=CC=C(C(=C1)CCBr)F
This item is not returnable. View return policy
91319-54-9
C8H8BrF
203.05
FQGDFWIQBCQXPS-UHFFFAOYSA-N
2-fluorophenethyl bromide, 1-2-bromoethyl-2-fluorobenzene, 1-2-bromo-ethyl-2-fluoro-benzene, benzene, 1-2-bromoethyl-2-fluoro, 2-2-bromoethyl-1-fluorobenzene, 1-2-bromoethyl-2-fluoro-benzene, 2-fluorophenyl-ethylbromide, acmc-209wk4, 2-fluorophenyl ethyl bromide, 2-2-fluorophenyl-ethylbromide
2756964
1-(2-bromoethyl)-2-fluorobenzene
C1=CC=C(C(=C1)CCBr)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 73°C to 75°C (20.0 Torr)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 203.05
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Fluorophenethyl bromide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Fluorophenethyl bromide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'