missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoroethanol, 95%, Thermo Scientific Chemicals

Catalog Number p-7043635
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
371-62-0
C2H5FO
64.059
MFCD00002828
GGDYAKVUZMZKRV-UHFFFAOYSA-N
ethanol, 2-fluoro, fluoroethanol, 2-fluoro-1-ethanol, ethylene fluorohydrin, beta-fluoroethanol, 2-fluoroethan-1-ol, monofluorethanol, monofluoroethanol, 2-fluoro-ethanol, unii-4m427a5hml
9737
2-fluoroethanol
C(CF)O
This item is not returnable. View return policy
371-62-0
C2H5FO
64.059
MFCD00002828
GGDYAKVUZMZKRV-UHFFFAOYSA-N
ethanol, 2-fluoro, fluoroethanol, 2-fluoro-1-ethanol, ethylene fluorohydrin, beta-fluoroethanol, 2-fluoroethan-1-ol, monofluorethanol, monofluoroethanol, 2-fluoro-ethanol, unii-4m427a5hml
9737
2-fluoroethanol
C(CF)O

2-Fluoroethanol, is used in the microwave spectra of two isotopic species of 2-fluoroethanol have been investigated. The Gg conformer of 2-fluoroethanol is isolated in low-temperature noble gas matrices. Single-photon IR photolysis of 2-fluoroethanol in solid argon has been investigated.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoroethanol, is used in the microwave spectra of two isotopic species of 2-fluoroethanol have been investigated. The Gg conformer of 2-fluoroethanol is isolated in low-temperature noble gas matrices. Single-photon IR photolysis of 2-fluoroethanol in solid argon has been investigated.

Solubility
Fully miscible.

Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point -27°C
Boiling Point 102°C to 104°C
Flash Point 31°C (87°F)
Quantity 1 g
UN Number UN3384
Beilstein 1730857
Formula Weight 64.06
Percent Purity 95%
Chemical Name or Material 2-Fluoroethanol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'