missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluorobenzyl alcohol, 98%, Thermo Scientific™
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Specifications
CAS | 446-51-5 |
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Color | Colorless to Yellow |
Density | 1.1700g/mL |
Boiling Point | 200.0°C |
Flash Point | 90°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 97.5% min. (GC) |
Packaging | Glass bottle |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'