missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzonitrile, 99%, ACROS Organics™

Brand:  Acros Organics 250051000

Code : 29

Additional Details : CAS Number : 394-47-8 Weight : 0.15000kg Transport : UN number : 3276 Chem class : 1 Pack group : III

Quantity 100g
Packaging Glass bottle

 View more versions of this product

Product Code. 10580394

  • £75.70 / 100g


EU Stock 1
Add to basket

Specifications

Chemical Identifiers

2-Fluorobenzonitrile
98.5
≥98.5%
C7H4FN
MFCD00001773
394-47-8
100.0
99%
121.12

Specifications

2-Fluorobenzonitrile
Clear colorless to light brownish
394-47-8
98.5
Glass bottle
Liquid
99%
121.12
1.1100g/mL
100g
Authentic
100.0
1.5063 to 1.5083 (20°C, 589nm)
≥98.5%
C7H4FN
MFCD00001773
Videos
SDS