missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzonitrile, 99%, ACROS Organics™

Brand:  Acros Organics 250050250

Code : 29

Additional Details : CAS Number : 394-47-8 Weight : 0.07500kg Transport : UN number : 3276 Chem class : 1 Pack group : III

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10481091

  • £26.70 / 25g


EU Stock 2
Add to basket

Specifications

Chemical Identifiers

2-Fluorobenzonitrile
98.5
≥98.5%
C7H4FN
MFCD00001773
394-47-8
100.0
99%
121.12

Specifications

2-Fluorobenzonitrile
Clear colorless to light brownish
394-47-8
98.5
Glass bottle
Liquid
99%
121.12
1.1100g/mL
25g
Authentic
100.0
1.5063 to 1.5083 (20°C, 589nm)
≥98.5%
C7H4FN
MFCD00001773
Videos
SDS