missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzoic acid, 99%, Thermo Scientific Chemicals

Catalog Number p-133948
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
445-29-4
C7H5FO2
140.11
MFCD00002405
NSTREUWFTAOOKS-UHFFFAOYSA-N
o-fluorobenzoic acid, benzoic acid, 2-fluoro, ortho-fluorobenzoic acid, o-fluorbenzoesaeure, benzoic acid, o-fluoro, fluorobenzoic acid, unii-64uz32koo4, o-fluorbenzoesaeure german, ofba, 2-fluoro-benzoic acid
9935
CHEBI:19577
2-fluorobenzoic acid
C1=CC=C(C(=C1)C(=O)O)F
This item is not returnable. View return policy
445-29-4
C7H5FO2
140.11
MFCD00002405
NSTREUWFTAOOKS-UHFFFAOYSA-N
o-fluorobenzoic acid, benzoic acid, 2-fluoro, ortho-fluorobenzoic acid, o-fluorbenzoesaeure, benzoic acid, o-fluoro, fluorobenzoic acid, unii-64uz32koo4, o-fluorbenzoesaeure german, ofba, 2-fluoro-benzoic acid
9935
CHEBI:19577
2-fluorobenzoic acid
C1=CC=C(C(=C1)C(=O)O)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 123°C to 125°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula FC6H4CO2H
Quantity 25 g
Beilstein 09, 333
Solubility Information Solubility in water: 7.2g/L (25°C)
Formula Weight 140.11
Percent Purity 99%
Physical Form Crystalline Powder or Needles
Chemical Name or Material 2-Fluorobenzoic acid, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'