missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Fluorobenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H61743.03

Code : NEW

Additional Details : CAS Number : 30058-40-3 Weight : 0.00100kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15438308

  • £19.80 / 1g


Estimated Shipment date
from Supplier 23-07-2018
Add to basket

Specifications

Chemical Identifiers

2-Fluorobenzenesulfonamide
175.18
UN2811
30058-40-3
MFCD00042283

Specifications

2-Fluorobenzenesulfonamide
1g
Solid
175.18
UN2811
162° to 166°C
30058-40-3
97%
MFCD00042283
Videos
SDS