missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzaldehyde, 97%, Thermo Scientific Chemicals

Catalog Number p-133930
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
446-52-6
C7H5FO
124.11
MFCD00003302
ZWDVQMVZZYIAHO-UHFFFAOYSA-N
o-fluorobenzaldehyde, benzaldehyde, 2-fluoro, benzaldehyde, o-fluoro, unii-7zx20dxb9z, 2-fluor-benzaldehyd, 2-fluoro-benzaldehyde, benzaldehyde, fluoro, ortho-fluorobenzaldehyde, 7zx20dxb9z, fluorobenzaldehyde
67970
2-fluorobenzaldehyde
C1=CC=C(C(=C1)C=O)F
This item is not returnable. View return policy
446-52-6
C7H5FO
124.11
MFCD00003302
ZWDVQMVZZYIAHO-UHFFFAOYSA-N
o-fluorobenzaldehyde, benzaldehyde, 2-fluoro, benzaldehyde, o-fluoro, unii-7zx20dxb9z, 2-fluor-benzaldehyd, 2-fluoro-benzaldehyde, benzaldehyde, fluoro, ortho-fluorobenzaldehyde, 7zx20dxb9z, fluorobenzaldehyde
67970
2-fluorobenzaldehyde
C1=CC=C(C(=C1)C=O)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point -44.5°C
Color Brown to Colorless
Density 1.1700g/mL
Boiling Point 172°C to 174°C
Flash Point 55°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula FC6H4CHO
Refractive Index 1.5205 to 1.5225
Quantity 25 mL
Beilstein 07, I, 132
Specific Gravity 1.17
Solubility Information Solubility in water: insoluble
Formula Weight 124.11
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Fluorobenzaldehyde, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Fluorobenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'