missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoroanisole, 98%, Thermo Scientific™

Catalog Number p-7082104
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
321-28-8
C7H7FO
126.13
MFCD00000316
JIXDOBAQOWOUPA-UHFFFAOYSA-N
2-fluoroanisole, o-fluoroanisole, benzene, 1-fluoro-2-methoxy, ortho-fluoroanisole, anisole, o-fluoro, 1-fluoro-2-methoxy-benzene, paragos 390201, timtec-bb sbb040818, fr bo1 wln, fluoroanisole
67576
1-fluoro-2-methoxybenzene
COC1=CC=CC=C1F
This item is not returnable. View return policy
321-28-8
C7H7FO
126.13
MFCD00000316
JIXDOBAQOWOUPA-UHFFFAOYSA-N
2-fluoroanisole, o-fluoroanisole, benzene, 1-fluoro-2-methoxy, ortho-fluoroanisole, anisole, o-fluoro, 1-fluoro-2-methoxy-benzene, paragos 390201, timtec-bb sbb040818, fr bo1 wln, fluoroanisole
67576
1-fluoro-2-methoxybenzene
COC1=CC=CC=C1F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point -39°C
Density 1.124
Boiling Point 154°C to 155°C
Flash Point 56°C (133°F)
Refractive Index 1.494
Quantity 25 g
UN Number UN1993
Beilstein 1859755
Formula Weight 126.13
Percent Purity 98%
Chemical Name or Material 2-Fluoroanisole
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'