2-Fluoroaniline, 99+%, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 119275000

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 348-54-9 Weight: 0.57950kg Transport: UN number : 1992 Chem class : 1 Pack group : III


Packaging Glass bottle
Quantity 500mL

Product Code. 10530584

Quantity Price
1 £ 95.1 / 500mL
Estimated Shipment date
from Supplier 23-05-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 2-Fluoroaniline
CAS 348-54-9
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0% (GC)
Assay Percent Range 99+%
Molecular Formula C6H6FN
Linear Formula FC6H4NH2
Formula Weight 111.12
MDL Number MFCD00007642

Specification

Assay ≥99.0% (GC)
Formula Weight 111.12
Physical Form Liquid
Quantity 500mL
Color Clear colorless
CAS Min % 99.0
Linear Formula FC6H4NH2
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
CAS 348-54-9
Assay Percent Range 99+%
MDL Number MFCD00007642
Refractive Index 1.5410 to 1.5430 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
Flammable liquid and vapor.
Packaging Glass bottle
Molecular Formula C6H6FN
Health Hazard 1 GHS Signal Word: Warning
CAS Max % 100.0
Density 1.151g/mL
Chemical Name or Material 2-Fluoroaniline
Beilstein 12, I, 296
Documentation

Technical documents

Safety and Handling

GHS Signal Word: Warning

GHS H Statement:
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.